Skip to Content
Merck
All Photos(2)

Key Documents

706086

Sigma-Aldrich

2,6-Dimethoxy-3-pyridineboronic acid

≥95%

Synonym(s):

(2,6-Dimethoxypyridin-3-yl)boronic acid, 2,6-Dimethoxypyridine-3-boronic acid

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€61.60

€61.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€61.60

About This Item

Empirical Formula (Hill Notation):
C7H10BNO4
CAS Number:
Molecular Weight:
182.97
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

€61.60


Check Cart for Availability

Request a Bulk Order

Assay

≥95%

form

powder or crystals

mp

100-117 °C

storage temp.

2-8°C

SMILES string

COc1ccc(B(O)O)c(OC)n1

InChI

1S/C7H10BNO4/c1-12-6-4-3-5(8(10)11)7(9-6)13-2/h3-4,10-11H,1-2H3

InChI key

ADGHSWFUZUADDH-UHFFFAOYSA-N

Application

May contain varying amounts of anhydride
Reactant for:
  • Preparation of benzopyranone derivatives as positive GABAA receptor modulators
  • Preparation of pyrazine derivatives as orally active corticotropin releasing factor-1 receptor antagonists
  • Microwave-enhanced synthesis of trisubstituted pyridazines
  • Preparation of biaryl derivatives via palladium-catalyzed Suzuki-Miyaura cross-coupling reaction with aryl- or heteroaryl halides
  • Preparation of 3-arylpyridines via palladium-P Phos catalyzed Suzuki cross coupling

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service