Skip to Content
Merck
All Photos(1)

Key Documents

705292

Sigma-Aldrich

Riboflavin-(dioxopyrimidine-13C4,15N2)

≥98 atom %, ≥97% (CP)

Synonym(s):

(–)-Riboflavin-(dioxopyrimidine-13C4,15N2), Lactoflavin-(dioxopyrimidine-13C4,15N2), Vitamin B2-(dioxopyrimidine-13C4,15N2), Vitamin G-(dioxopyrimidine-13C4,15N2)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€316.00
5 MG
€1,230.00

€316.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 MG
€316.00
5 MG
€1,230.00

About This Item

Empirical Formula (Hill Notation):
13C4C13H2015N2N2O6
CAS Number:
Molecular Weight:
382.32
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.24

€316.00


Check Cart for Availability

Request more information

isotopic purity

 13C
 15N
≥98 atom %

Quality Level

Assay

≥97% (CP)

form

chunks

technique(s)

mass spectrometry (MS): suitable

color

orange

mp

290 °C (dec.) (lit.)

mass shift

M+6

storage temp.

−20°C

SMILES string

Cc1cc2N=[13C]3[13C](=O)[15NH][13C](=O)[15N]=[13C]3N(C[C@H](O)[C@H](O)[C@H](O)CO)c2cc1C

InChI

1S/C17H20N4O6/c1-7-3-9-10(4-8(7)2)21(5-11(23)14(25)12(24)6-22)15-13(18-9)16(26)20-17(27)19-15/h3-4,11-12,14,22-25H,5-6H2,1-2H3,(H,20,26,27)/t11-,12+,14-/m0/s1/i13+1,15+1,16+1,17+1,19+1,20+1

InChI key

AUNGANRZJHBGPY-HAIGJTSVSA-N

Related Categories

General description

Riboflavin (Vitamin B2) is found in certain foods such as milk, meat, eggs, nuts, enriched flour, and green vegetables.[1][2] It is necessary for normal cell growth and function.

Application

Riboflavin-(dioxopyrimidine-13C4,15N2) can be used as an analytical standard for the quantitative analysis of riboflavin content in human plasma via LC-MS/MS.[3]

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Riboflavin
Pinto JT and Zempleni J
Advances in Nutrition (Bethesda, Md.), 7(5), 973-975 (2016)
Isotope dilution method for determination of vitamin B2 in human plasma using liquid chromatography-tandem mass spectrometry
Diniz, M, et al.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 1113(5), 14-19 (2019)
Riboflavin (vitamin B-2) and health
Powers Hilary J
American Journal of Clinical Nutrition, 77(6), 1352-1360 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service