Skip to Content
Merck
All Photos(2)

Key Documents

702307

Sigma-Aldrich

5-Bromo-3-iodo-7-azaindole

97%

Synonym(s):

5-Bromo-3-iodo-1H-pyrrolo[2,3-b]pyridine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€162.00

€162.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€162.00

About This Item

Empirical Formula (Hill Notation):
C7H4BrIN2
CAS Number:
Molecular Weight:
322.93
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€162.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

powder

mp

234-239 °C

functional group

bromo
iodo

SMILES string

Brc1cnc2[nH]cc(I)c2c1

InChI

1S/C7H4BrIN2/c8-4-1-5-6(9)3-11-7(5)10-2-4/h1-3H,(H,10,11)

InChI key

GIPGJYARDOQGDJ-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pavel Štarha et al.
PloS one, 10(8), e0136338-e0136338 (2015-08-27)
The cis-[PtCl2(naza)2] complexes (1-3) containing monosubstituted 7-azaindole halogeno-derivatives (naza), showed significantly higher activity than cisplatin towards ovarian carcinoma A2780, its cisplatin-resistant variant A2780R, osteosarcoma HOS, breast carcinoma MCF7 and cervix carcinoma HeLa cell lines, with the IC50 values of 3.8

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service