Skip to Content
Merck
All Photos(2)

Key Documents

702293

Sigma-Aldrich

5-Bromo-1H-indazole

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€50.50

€50.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€50.50

About This Item

Empirical Formula (Hill Notation):
C7H5BrN2
CAS Number:
Molecular Weight:
197.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€50.50


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

powder

mp

123-127 °C

functional group

bromo

SMILES string

Brc1ccc2[nH]ncc2c1

InChI

1S/C7H5BrN2/c8-6-1-2-7-5(3-6)4-9-10-7/h1-4H,(H,9,10)

InChI key

STVHMYNPQCLUNJ-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
568279666521734489
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

form

powder

form

solid

form

solid

form

solid

mp

123-127 °C

mp

40-44 °C (lit.)

mp

197-202 °C

mp

150-155 °C

functional group

bromo

functional group

bromo

functional group

bromo

functional group

bromo, ketone

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service