700916
12-Bromododecanonitrile
97%
Synonym(s):
11-Bromoundecyl cyanide, 12-Bromododecanenitrile, 12-Bromolauronitrile
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C12H22BrN
CAS Number:
Molecular Weight:
260.21
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Assay
97%
refractive index
n20/D 1.473
density
1.110 g/mL at 25 °C
SMILES string
BrCCCCCCCCCCCC#N
InChI
1S/C12H22BrN/c13-11-9-7-5-3-1-2-4-6-8-10-12-14/h1-11H2
InChI key
REWUNQPQBYXSQB-UHFFFAOYSA-N
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
>230.0 °F
Flash Point(C)
> 110 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service