Skip to Content
Merck
All Photos(1)

Key Documents

700177

Sigma-Aldrich

Indium oxide coated PET

surface resistivity ≤10 Ω/sq, slide

Synonym(s):

IO-PET, Polyethylene terephthalate film, indium oxide coated

Sign Into View Organizational & Contract Pricing

Select a Size

5 PKG
€766.00

€766.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 PKG
€766.00

About This Item

Empirical Formula (Hill Notation):
In2O3
Molecular Weight:
277.63
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

€766.00


Check Cart for Availability

Request a Bulk Order

Quality Level

form

solid

surface resistivity

≤10 Ω/sq

L × W × thickness

150 × 150 × 0.2 mm

SMILES string

O=[In]O[In]=O

InChI

1S/2In.3O

InChI key

SHTGRZNPWBITMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

In2O3/Au/Ag coated PET film

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service