Skip to Content
Merck
All Photos(1)

Key Documents

700037

Sigma-Aldrich

(Ethylenediamine)iodoplatinum(II) dimer dinitrate

96%

Synonym(s):

Bis(1,2-ethanediamine-N,N′)di-μ-iododiplatinum(II) dinitrate, Di-μ-iodobis(ethylenediamine)diplatinum(II) dinitrate

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€189.00

€189.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€189.00

About This Item

Linear Formula:
C4H16I2N4Pt2 · 2NO3
CAS Number:
Molecular Weight:
888.17
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

€189.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

96%

form

solid

reaction suitability

core: platinum
reagent type: catalyst

mp

268 °C

SMILES string

I[Pt+].I[Pt+].NCCN.NCCN.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/2C2H8N2.2HI.2NO3.2Pt/c2*3-1-2-4;;;2*2-1(3)4;;/h2*1-4H2;2*1H;;;;/q;;;;2*-1;2*+2/p-2

InChI key

NRXMBGQJNZDBOD-UHFFFAOYSA-L

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T V O'Halloran et al.
Journal of molecular biology, 194(4), 705-712 (1987-04-20)
The X-ray structure of the nucleosome core particle was solved at 7 A resolution using the method of multiple isomorphous replacement based on two isomorphous derivatives, each containing a different multiple heavy-atom compound. The preparation of these heavy-atom compounds and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service