Skip to Content
Merck
All Photos(3)

Key Documents

698695

Sigma-Aldrich

Carbon nanotube, single-walled

>70 wt. % carbon basis, D × L 2-10 nm × 1-5 μm , bundle dimensions, avg. no. of layers, 1

Synonym(s):

SWCNT, SWNT, Single wall carbon nanotube

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

Assay

>70% carbon basis (wt.)

feature

avg. no. of layers 1

composition

carbon, ≥70%

D × L

2-10 nm × 1-5 μm , bundle dimensions

size

1.3-1.5 nm , individual SWNT diameter

mp

3652-3697 °C (lit.)

density

1.7-1.9 g/cm3 at 25 °C (lit.)

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Carbon nanotube, single-walled (SWNT) belongs to the class of carbonaceous materials with excellent physiochemical, thermo-mechanical and electrochemical properties. This material can be used in a variety of sustainable energy applications such as solar cells, photocatalysis, thin film conductors, field effect transistors (FETs), biosensor, gas sensor, supercapacitor and nanomechanical resonators.[1][2][3][4][5][6][7]
Metal impurities in single walled carbon nanotubes (SWNTs) were studied as airborne tracers of carbon nanotubes (CNTs) for workplace safety monitoring.[8] SWNTs were used as reinforcements in crosslinked composites of polypropylene fumarate (PPF) for bone tissue engineering applications.[9]

Preparation Note

Electric Arc Discharge Method

Analysis Note

Carbonaceous content[10]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Two-Dimensional Nanostructure- Reinforced Biodegradable
Polymeric Nanocomposites for Bone Tissue Engineering
Lalwani G, et al.
Biomacromolecules, 14(3), 900-909 (2013)
Rasmussen PE, et al.
Journal of Physics. Conference Series, 429 (2013)
Catherine Kanimozhi et al.
ACS applied materials & interfaces, 9(46), 40734-40742 (2017-10-27)
Separation of electronically pure, narrowly dispersed, pristine, semiconducting single-walled carbon nanotubes (CNTs) from a heterogeneous as-synthesized mixture is essential for various semiconducting technologies and biomedical applications. Although conjugated polymer wrappers are often utilized to facilitate electronic-type sorting, it is highly
Electrochemical behaviour of different redox probes on single wall
carbon nanotube buckypaper-modified electrodes
Sieben JM, et al.
Electrochimica Acta, 135, 404-411c-404-411c (2014)
Single-walled carbon nanotubes as ultrahigh frequency nanomechanical resonators
Li C and Chou T
Physical Review. B, Condensed Matter and Materials Physics, 68(7), 073405-073405 (2003)

Articles

Single-Walled Carbon Nanotubes synthesized by the Super-Growth Method & their properties & applications, including dispersing SGCNTs, SGCNT-polymer composites & SGCNT-metal composites are discussed.

Carbon nanotubes are materials that possess remarkable properties and offer extraordinary possibilities.

Carbon nanotubes (CNTs) have received much attention since their discovery in 1991 by Sumio lijima1 due to their excellent mechanical, electrical, and optical properties.

A nanocomposite is typically defined as a mixture between a host material (e.g., polymer matrix) and nanofillers with at least one dimension of less than 100 nm.

See All

Protocols

Surfactant-assisted dispersion of single-walled carbon nanotubes for debundling or exfoliation in dispersion procedures.

Questions

1–2 of 2 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. What is the chirality of product 698695, Carbon nanotube, single-walled?

    1 answer
    1. Product 698695, Carbon nanotube, consists of as-produced electric arc nanotubes with various chiralities (n, m values), and those individual chiralities have not been seperated for this product listing.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service