Skip to Content
Merck
All Photos(1)

Key Documents

698385

Sigma-Aldrich

1,2-Bis[(2S,5S)-2,5-diethylphospholano]benzene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate

Synonym(s):

(S,S)-Et-DUPHOS-Rh

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€238.00

€238.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€238.00

About This Item

Empirical Formula (Hill Notation):
C30H48BF4P2Rh
CAS Number:
Molecular Weight:
660.36
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

€238.00


Check Cart for Availability

Request a Bulk Order

form

powder

functional group

phosphine

SMILES string

[Rh+].F[B-](F)(F)F.C1CC=CCCC=C1.CC[C@H]2CC[C@H](CC)P2c3ccccc3P4[C@@H](CC)CC[C@@H]4CC

InChI

1S/C22H36P2.C8H12.BF4.Rh/c1-5-17-13-14-18(6-2)23(17)21-11-9-10-12-22(21)24-19(7-3)15-16-20(24)8-4;1-2-4-6-8-7-5-3-1;2-1(3,4)5;/h9-12,17-20H,5-8,13-16H2,1-4H3;1-2,7-8H,3-6H2;;/q;;-1;+1/b;2-1-,8-7-;;/t17-,18-,19-,20-;;;/m0.../s1

InChI key

DSYBEQKPSKLNMC-ZCTOJWETSA-N

Application

DuPhos and BPE Ligands: Highly Efficient Privileged Ligands

Catalyst for:
  • Preparation of a key unit B precursor of cryptophycins via Horner-Wadsworth-Emmons reaction and asymmetric hydrogenation
  • Preparation of constrained phenylalanine analogs via Heck reaction followed by asymmetric hydrogenation and cyclization steps
  • Enantioselective preparation of N-benzyloxy-β-amino acid Me esters by hydrogenation of α-(benzyloxyaminomethyl)acrylate stereoisomers
  • Preparation of fluoro amino acids as synthons for potent macrocyclic HCV NS3 protease inhibitors
  • Stereoselective preparation of triply isotope-labeled Ser, Cys, and Ala
  • Preparation of β-amino acids from asymmetric hydrogenation of α-(aminomethyl)acrylates
  • Asymmetric hydrogenation of a-primary and secondary amino ketones and asymmetric synthesis of (-)-arbutamine and (-)-denopamine

Legal Information

Sold in collaboration with Kanata Chemical Technologies Inc. for research purposes only. These compounds were made and sold under license from E.I. du Pont de Nemours and Company, which license does not include the right to use the compounds in producing products for sale in the pharmaceutical field.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service