Skip to Content
Merck
All Photos(1)

Key Documents

698202

Sigma-Aldrich

2,5-Bis(1-naphthyl)-1,3,4-oxadiazole

99%

Synonym(s):

2,5-Di(naphthalen-1-yl)-1,3,4-oxadiazole, BND

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H14N2O
CAS Number:
Molecular Weight:
322.36
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

solid

mp

175-178 °C

SMILES string

c1ccc2c(cccc2c1)-c3nnc(o3)-c4cccc5ccccc45

InChI

1S/C22H14N2O/c1-3-11-17-15(7-1)9-5-13-19(17)21-23-24-22(25-21)20-14-6-10-16-8-2-4-12-18(16)20/h1-14H

InChI key

MUNFOTHAFHGRIM-UHFFFAOYSA-N

Application

Organic electronic material useful as an electron transporter in organic light emitting diodes (OLEDs).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tareq Hameduh et al.
Computational and structural biotechnology journal, 18, 3494-3506 (2020-12-12)
Homology modeling is a method for building protein 3D structures using protein primary sequence and utilizing prior knowledge gained from structural similarities with other proteins. The homology modeling process is done in sequential steps where sequence/structure alignment is optimized, then
H. Tokuhisa, M. Era, T. Tsutsui, S. Saito.
Applied Physics Letters, 25, 3433-3433 (1995)
S. Hoshino, K. Ebata, K. Furukawa.
Journal of Applied Physics, 4, 2000-2000 (1968)
J Andrew Duty et al.
The Journal of experimental medicine, 206(1), 139-151 (2008-12-24)
Self-reactive B cells not controlled by receptor editing or clonal deletion may become anergic. We report that fully mature human B cells negative for surface IgM and retaining only IgD are autoreactive and functionally attenuated (referred to as naive IgD(+)IgM(-)
Elisabeth Ott et al.
PloS one, 6(5), e18609-e18609 (2011-05-24)
OriP, the latent origin of Epstein-Barr virus (EBV), consists of two essential elements: the dyad symmetry (DS) and the family of repeats (FR). The function of these elements has been predominantly analyzed in plasmids transfected into transformed cells. Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service