Skip to Content
Merck
All Photos(2)

Key Documents

698008

Sigma-Aldrich

3-Hydroxyphenylboronic acid MIDA ester

97%

Synonym(s):

2-(3-Hydroxyphenyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12BNO5
CAS Number:
Molecular Weight:
249.03
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

204-209 °C

SMILES string

CN1CC(=O)OB(OC(=O)C1)c2cccc(O)c2

InChI

1S/C11H12BNO5/c1-13-6-10(15)17-12(18-11(16)7-13)8-3-2-4-9(14)5-8/h2-5,14H,6-7H2,1H3

InChI key

NZNGUMOTSLXTJB-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service