Skip to Content
Merck
All Photos(2)

Key Documents

697109

Sigma-Aldrich

3-Fluoro-2-pyridinecarbonitrile

96%

Synonym(s):

2-Cyano-3-fluoropyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3FN2
CAS Number:
Molecular Weight:
122.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

96%

form

crystals

mp

27-32 °C

functional group

fluoro
nitrile

SMILES string

Fc1cccnc1C#N

InChI

1S/C6H3FN2/c7-5-2-1-3-9-6(5)4-8/h1-3H

InChI key

VZFPSCNTFBJZHB-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

219.9 °F

Flash Point(C)

104.4 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service