Skip to Content
Merck
All Photos(1)

Key Documents

696684

Sigma-Aldrich

2-Chloro-3-quinolineboronic acid

≥95%

Synonym(s):

(2-Chloro-3-quinolinyl)boronic acid, 2-Chloroquinolin-3-ylboronic acid, 2-Chloroquinoline-3-boronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7BClNO2
CAS Number:
Molecular Weight:
207.42
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

≥95%

form

solid

storage temp.

2-8°C

SMILES string

OB(O)c1cc2ccccc2nc1Cl

InChI

1S/C9H7BClNO2/c11-9-7(10(13)14)5-6-3-1-2-4-8(6)12-9/h1-5,13-14H

InChI key

JAQXYUOPSOXQCG-UHFFFAOYSA-N

Application

May contain varying amounts of anhydride
Reactant for:
  • Preparation of pyrazolopyrimidinamine derivatives as inhibitors of tyrosine and phosphinositide kinase

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service