Skip to Content
Merck
All Photos(1)

Key Documents

693715

Sigma-Aldrich

Terbium

foil, size 25 mm × 25 mm × 1 mm, 99.9% trace rare earth metals basis

Synonym(s):

Terbium-159

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Tb
CAS Number:
Molecular Weight:
158.93
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:

Assay

99.9% trace rare earth metals basis

form

foil

reaction suitability

reagent type: catalyst
core: terbium

resistivity

116 μΩ-cm, 20°C

size

25 mm × 25 mm × 1 mm

bp

3230 °C (lit.)

mp

1356 °C (lit.)

density

8.234 g/mL at 25 °C (lit.)

SMILES string

[Tb]

InChI

1S/Tb

InChI key

GZCRRIHWUXGPOV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in Physical Vapor Deposition (PVD) processes, including thermal and electron-beam (e-beam) evaporation, for the preparation of thin films.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junfeng Hui et al.
Nanoscale, 4(22), 6967-6970 (2012-10-19)
Water-dispersible hydrophilic fluoridated HAp:Ln(3+) (Ln = Eu or Tb) nanoparticles were prepared via hydrophobic/hydrophilic transformation with surfactants (Pluronic F127). The HAp:Ln(3+) (Ln = Eu or Tb) nanoparticles with unique luminescent properties and excellent biocompatibility are promising for cell imaging applications.
Hongyu Chen et al.
ACS nano, 7(2), 1178-1187 (2013-01-04)
One of the greatest challenges in cancer therapy is to develop methods to deliver chemotherapy agents to tumor cells while reducing systemic toxicity to noncancerous cells. A promising approach to localizing drug release is to employ drug-loaded nanoparticles with coatings
Xi Juan Zhao et al.
The Analyst, 137(22), 5190-5192 (2012-10-04)
Highly selective sensing of cytidine triphosphate (CTP) against other triphosphate nucleosides including ATP, GTP and UTP is successfully achieved with a luminescent terbium(III)-organic framework (TbOF) of [Tb(2)(2,3-pzdc)(2)(ox)(H(2)O)(2)](n) (2,3-pzdc(2-) = 2,3-pyrazinedicarboxylate, ox(2-) = oxalate).
Jing Zhang et al.
Biosensors & bioelectronics, 41, 704-709 (2012-10-24)
On the basis of Tb(3+), structure-switching aptamer and magnetic beads (MBs), a signal-on fluorescent aptasensor was developed for the label-free determination of OTA in wheat. Initially, the specific sequence of the anti-OTA aptamer labeled with a biotin group, was attached
Swagatika Sahu et al.
Dalton transactions (Cambridge, England : 2003), 42(6), 2224-2231 (2012-12-01)
In this paper fluorescent magnetic Fe(3)O(4)@m-SiO(2)@YPO(4):Tb(3+) particles with a size of 52 nm have been prepared and characterized. The surface of the nanoparticles was modified with β-cyclodextrin and folic acid using glutathione as a linker. The hydrophobic anticancer drug 5-fluorouracil

Articles

A significant limiting factor for wearable electronics and wireless sensors is the finite amount of energy that can be stored in on-board batteries.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service