Skip to Content
Merck
All Photos(1)

Key Documents

693677

Sigma-Aldrich

Thulium

foil, size 25 mm × 25 mm × 1 mm, 99.9% trace rare earth metals basis

Synonym(s):

Thulium-169

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Tm
CAS Number:
Molecular Weight:
168.93
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace rare earth metals basis

form

foil

reaction suitability

reagent type: catalyst
core: thulium

resistivity

90 μΩ-cm, 20°C

size

25 mm × 25 mm × 1 mm

bp

1950 °C (lit.)

mp

1545 °C (lit.)

density

9.332 g/mL at 25 °C (lit.)

SMILES string

[Tm]

InChI

1S/Tm

InChI key

FRNOGLGSGLTDKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in Physical Vapor Deposition (PVD) processes, including thermal and electron-beam (e-beam) evaporation, for the preparation of thin films.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhonghua Yang et al.
Urology, 81(2), 396-400 (2013-02-05)
To compare the clinical outcomes between thulium laser transurethral enucleation of the prostate (ThuLEP) and plasmakinetic bipolar resection of the prostate (PKRP) for treating benign prostatic hyperplasia (BPH) in a prospective randomized trial with 18 months of follow-up. The study
Commentary: A split-face study using the 1,927-nm thulium fiber fractional laser for photoaging and melasma in Asian skin.
Kristel D Polder
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.], 39(6), 889-890 (2013-06-07)
Bo Peng et al.
BJU international, 111(4), 633-637 (2012-10-31)
WHAT'S KNOWN ON THE SUBJECT? AND WHAT DOES THE STUDY ADD?: Thulium laser is a new generation of surgical laser. It is a minimally invasive technology with several advantages, including rapid vaporization and minimal tissue damage and bleeding. However, details
Jacek Swiderski et al.
Optics express, 21(7), 7851-7857 (2013-04-11)
We demonstrate a novel method of mid-infrared (mid-IR) supercontinuum (SC) generation with the use of a 2 µm gain-switched self-mode-locked thulium-doped fiber laser. SC radiation ranging from ~1.9 to 3.8 µm wavelength, generated in a single-mode ZBLAN fiber with a
Rajesh Kadel et al.
Applied optics, 51(27), 6465-6470 (2012-10-04)
We have demonstrated a self-starting, passively mode-locked Tm/Ho codoped fiber laser that lases at one of two center wavelengths. An amplified 1.56 μm distributed feedback laser pumps a ring laser cavity which contains 1 m of Tm/Ho codoped silica fiber.

Articles

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service