Skip to Content
Merck
All Photos(2)

Key Documents

693510

Sigma-Aldrich

Gadolinium-silicon-germanium alloy

Gd5Si2Ge2, ≥99% trace metals basis

Synonym(s):

Gadolinium germanium silicide

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€102.00

€102.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€102.00

About This Item

Linear Formula:
Gd5Ge2Si2
CAS Number:
Molecular Weight:
987.70
MDL number:
UNSPSC Code:
11101711
PubChem Substance ID:
NACRES:
NA.23

€102.00


Check Cart for Availability

Request a Bulk Order

Assay

≥99% trace metals basis

form

powder and chunks

composition

Gadolinium, 79.5 wt. %
Germanium, 14.8 wt. %
Silicon, 5.7 wt. %

mp

1675 °C

density

5.7 g/mL at 25 °C

SMILES string

[Si].[Ge].[Gd]

InChI

1S/Gd.Ge.Si

InChI key

LVKVKSQVTJLPDE-UHFFFAOYSA-N

Application

This material is used for research in the area of magnetic refrigeration. Alloy with a giant magnetocaloric effect.

Physical properties

Curie Temperature: ~270 K

Legal Information

Sold under license to US Patents 5,887,449 and 5,743,095 for use in magnetic refrigeration.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Water-react 3

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Today, near-room-temperature refrigeration is almost entirely based on a vapor-compression refrigeration cycle.

Solid state and materials chemistry have made a tremendous impact and have experienced growth in recent years, particularly for rare earthcontaining materials.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service