Skip to Content
Merck
All Photos(1)

Key Documents

691658

Sigma-Aldrich

Iron phosphide, Fe2P

99.5% trace metals basis

Synonym(s):

Ferrous phosphide, Iron diphosphide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe2P
CAS Number:
Molecular Weight:
142.66
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

99.5% trace metals basis

form

powder

particle size

−40 mesh

SMILES string

[Fe]=P#[Fe]

InChI

1S/2Fe.P

InChI key

GNTCPDPNMAMZFW-UHFFFAOYSA-N

General description

Iron phosphide, Fe2P is a low band semiconducting material. It can be prepared by sonochemical synthesis from organometallic compounds as starting materials at temperatures above 950°C.[1]

Application

Fe2P can be used for a variety of application such as: lithium ion batteries to improve conductivity[2], electrode material in sodium ion batteries[3], an electrocatalyst in hydrogen evolution[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhanced photocatalytic hydrogen production in water under visible light using noble metal-free ferrous phosphide as an active cocatalyst
Sun Z, et al.
Catalysis Science & Technology, 5(11), 4964-4967 (2015)
Iron phosphide as negative electrode material for Na-ion batteries
Zhang W, et al.
Electrochemical Communications, 69(11), 11-14 (2016)
A solvothermal synthesis of ultra-fine iron phosphide
Yunle G, et al.
Materials Research Bulletin, 37(6), 1101-1105 (2002)
Electrochemical activity of iron phosphide nanoparticles in hydrogen evolution reaction
Tian L, et al.
ACS Catalysis, 6(8), 5441-5448 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service