Skip to Content
Merck
All Photos(1)

Key Documents

690422

Sigma-Aldrich

2-Bromocyclohexanone

≥90% (GC)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€172.00

€172.00


Estimated to ship on17 April 2025


Request a Bulk Order

Select a Size

Change View
1 G
€172.00

About This Item

Empirical Formula (Hill Notation):
C6H9BrO
CAS Number:
Molecular Weight:
177.04
Beilstein:
1071730
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€172.00


Estimated to ship on17 April 2025


Request a Bulk Order

Quality Level

Assay

≥90% (GC)

form

liquid

refractive index

n20/D 1.512

functional group

bromo
ketone

storage temp.

−20°C

SMILES string

BrC1CCCCC1=O

InChI

1S/C6H9BrO/c7-5-3-1-2-4-6(5)8/h5H,1-4H2

InChI key

KDXYEWRAWRZXFT-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service