Skip to Content
Merck
All Photos(1)

Key Documents

690228

Sigma-Aldrich

N,N-Bis[3-(methylamino)propyl]methylamine

98%

Synonym(s):

N-Methyl-3,3′-bis(methylamino)dipropylamine, N-Methyl-N,N-bis[3-(methylamino)propyl]amine, Methylbis(3-methylaminopropyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3N(CH2CH2CH2NHCH3)2
CAS Number:
Molecular Weight:
173.30
Beilstein:
6711228
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.5% (GC)
97.5-102.5% (with HClO4, T)
98%

form

liquid

impurities

≤1% water

SMILES string

CNCCCN(C)CCCNC

InChI

1S/C9H23N3/c1-10-6-4-8-12(3)9-5-7-11-2/h10-11H,4-9H2,1-3H3

InChI key

PQYGBJHVVYREGU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Nochlin et al.
Journal of neuropathology and experimental neurology, 41(2), 233-240 (1982-03-01)
The administration of an aliphatic triamine, 3,3'-methylimino-bis-(N-methylpropylamine), was used to produce non-necrotizing lesions in the hypothalamus and medulla oblongata of rats. The distribution of the lesions in the hypothalamus was similar to that produced by goldthioglucose, and consisted ultrastructurally of
Gold thioglucose, mimpa and lesions of the ventromedial hypothalamus.
D F Brown
Journal of neuropathology and experimental neurology, 43(4), 455-457 (1984-07-01)
S Levine et al.
Journal of immunological methods, 54(3), 355-360 (1982-11-12)
In the past, the lesions of experimental allergic encephalomyelitis (EAE) have been induced to localize around brain tissue damaged by anoxia or direct physical or chemical attack. The procedure for producing the requisite antecedent brain injury has been simplified by
D F Brown et al.
Physiology & behavior, 49(1), 41-46 (1991-01-01)
It has been demonstrated that the ventromedial hypothalamus (VMH) of alloxan-induced diabetic mice is protected from subsequent gold thioglucose (GTG)-induced lesions. Another compound, 3,3'-methyliminobis-(N-methylpropylamine) (MIMPA), a triamine structurally unrelated to GTG, has been shown to cause similar VMH lesions in
S Levine et al.
Brain research, 242(2), 219-225 (1982-06-24)
An aliphatic triamine has been reported to cause lesions in rats in the vicinity of the area postrema and the median eminence of the hypothalamus, sites known to lack a blood-brain barrier. The present study revealed that some of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service