Skip to Content
Merck
All Photos(1)

Documents

689955

Sigma-Aldrich

1-Bromo-4,5-dimethoxy-2-nitrobenzene

99%

Synonym(s):

2-Bromo-4,5-dimethoxynitrobenzene, 4-Bromo-5-nitroveratrole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8BrNO4
CAS Number:
Molecular Weight:
262.06
Beilstein:
2270367
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (HPLC)
99%

form

powder

SMILES string

COc1cc(Br)c(cc1OC)[N+]([O-])=O

InChI

1S/C8H8BrNO4/c1-13-7-3-5(9)6(10(11)12)4-8(7)14-2/h3-4H,1-2H3

InChI key

DGUDEQARXVYDBS-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service