Skip to Content
Merck
All Photos(1)

Key Documents

689866

Sigma-Aldrich

Benzyl acetoacetate

Arxada quality, ≥96.0% (GC)

Synonym(s):

Acetoacetic acid benzyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2COOCH2C6H5
CAS Number:
Molecular Weight:
192.21
Beilstein:
2049615
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Pricing and availability is not currently available.

Assay

≥96.0% (GC)

form

liquid

quality

Arxada quality

manufacturer/tradename

Arxada AG

impurities

benzylic alcohol
≤0.05% water

refractive index

n20/D 1.512 (lit.)

bp

156-159 °C/10 mmHg (lit.)

density

1.112 g/mL at 25 °C (lit.)

SMILES string

CC(=O)CC(=O)OCc1ccccc1

InChI

1S/C11H12O3/c1-9(12)7-11(13)14-8-10-5-3-2-4-6-10/h2-6H,7-8H2,1H3

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service