Skip to Content
Merck
All Photos(2)

Key Documents

68972

Sigma-Aldrich

Tri-Boc-hydrazinoacetic acid

≥97.0% (N), for peptide synthesis

Synonym(s):

Tri-tert-butyl N-(carboxymethyl)hydrazinetricarboxylate, tri-Boc-hydrazinoacetic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€246.00

€246.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€246.00

About This Item

Empirical Formula (Hill Notation):
C17H30N2O8
CAS Number:
Molecular Weight:
390.43
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

€246.00


Check Cart for Availability

Request a Bulk Order

Product Name

Tri-Boc-hydrazinoacetic acid, ≥97.0% (N)

Assay

≥97.0% (N)

form

powder

reaction suitability

reagent type: ligand

mp

112-115 °C

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)N(CC(O)=O)N(C(=O)OC(C)(C)C)C(=O)OC(C)(C)C

InChI

1S/C17H30N2O8/c1-15(2,3)25-12(22)18(10-11(20)21)19(13(23)26-16(4,5)6)14(24)27-17(7,8)9/h10H2,1-9H3,(H,20,21)

InChI key

FSRNHDLQBNOYJK-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Peeyush Kumar Sharma et al.
ACS applied materials & interfaces, 10(37), 30936-30945 (2018-08-28)
Polymeric hydrogels have been extensively explored for controlled drug-delivery applications, but there is an increasing demand for smart drug delivery combined with tunable physicochemical attributes and tissue engineering potential. In this work, novel xanthan-poly(ethylene glycol) (PEG) hydrogels were developed by
Peeyush K Sharma et al.
Biomacromolecules, 20(6), 2174-2184 (2019-04-26)
Precise local delivery of chemotherapeutic agents employing an injectable depot could be a promising approach to achieve spatiotemporal control over the drug release along with minimizing the challenges associated with the systemic delivery of chemotherapeutic agents. In this regard, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service