Skip to Content
Merck
All Photos(3)

Key Documents

68794

Sigma-Aldrich

4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-Heptadecafluoroundecyl iodide

≥98%

Synonym(s):

1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-Heptadecafluoro-11-iodoundecane, 1H,1H,2H,2H,3H,3H-Perfluoroundecyl iodide, 3-(Perfluorooctyl)propyl iodide

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€70.30
5 G
€268.00

€70.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€70.30
5 G
€268.00

About This Item

Empirical Formula (Hill Notation):
C11H6F17I
CAS Number:
Molecular Weight:
588.04
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

€70.30


Check Cart for Availability

Request a Bulk Order

Assay

≥98%

form

powder

mp

32-36 °C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCCI

InChI

1S/C11H6F17I/c12-4(13,2-1-3-29)5(14,15)6(16,17)7(18,19)8(20,21)9(22,23)10(24,25)11(26,27)28/h1-3H2

InChI key

AXUBYTAXJHIPJO-UHFFFAOYSA-N

Related Categories

Legal Information

Use of this compound may be protected by U.S. patents 6,156,896; 5,859,247; 5,777,121 and 6,673,539.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - STOT RE 1

Target Organs

Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service