Skip to Content
Merck
All Photos(1)

Key Documents

687359

Sigma-Aldrich

Fmoc-Asn-OH-13C4,15N2

98 atom % 15N, 99 atom % 13C, 95% (CP)

Synonym(s):

Fmoc-L-Asparagine-13C4,15N2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C4C15H1815N2O5
Molecular Weight:
360.31
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
Pricing and availability is not currently available.

isotopic purity

99 atom % 13C
98 atom % 15N

Assay

95% (CP)

mp

~190 °C (lit.)

application(s)

peptide synthesis

functional group

Fmoc

mass shift

M+6

storage temp.

2-8°C
2-8°C

SMILES string

[15NH2][13C](=O)[13CH2][13C@H]([15NH]C(=O)OCC1c2ccccc2-c3ccccc13)[13C](O)=O

InChI

1S/C19H18N2O5/c20-17(22)9-16(18(23)24)21-19(25)26-10-15-13-7-3-1-5-11(13)12-6-2-4-8-14(12)15/h1-8,15-16H,9-10H2,(H2,20,22)(H,21,25)(H,23,24)/t16-/m0/s1/i9+1,16+1,17+1,18+1,20+1,21+1

InChI key

YUGBZNJSGOBFOV-LMDSVMOMSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service