Skip to Content
Merck
All Photos(1)

Key Documents

687316

Sigma-Aldrich

PEDOT

bis-PEG, lauryl terminated, 0.7 wt. % (nitromethane dispersion)

Synonym(s):

Aedotron C3-NM, C12-PEG-block-PEDOT-block-PEG-C12, PEDOT:PEG

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€295.00

€295.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€295.00

About This Item

CAS Number:
UNSPSC Code:
12352103
NACRES:
NA.23

€295.00


Check Cart for Availability

Request a Bulk Order

Product Name

Poly(3,4-ethylenedioxythiophene), bis-poly(ethyleneglycol), lauryl terminated, 0.7 wt. % (dispersion in nitromethane)

form

liquid (dispersion)

Quality Level

contains

perchlorate as dopant

composition

Acetonitrile, 4-8 wt. %
Aedotron- C3 polymer, 0.2-0.7 wt. %
Nitromethane, 90-95 wt. %
proprietary processing additive, 0.1-0.7 wt. %
Propylene glycol, 0.0-0.3 wt. %

concentration

0.5-0.9 wt. % (content of dispersion)
0.7 wt. % (dispersion in nitromethane)

surface resistivity

600-3000 Ω/sq

transmittance

70-85%

particle size

200-600 nm

conductivity

10-60 S/cm

Looking for similar products? Visit Product Comparison Guide

General description

Aedotron polymers are solvent-dispersable forms of PEDOT. These are copolymers of doped PEDOT and a flexible, soluble polymer such as poly(ethylene glycol) (PEG). These colloidal dispersions are stabilized by the highly solvated PEG chains which sterically limit the aggregation of the PEDOT blocks. Aedotron polymers are neither acidic nor corrosive, it easily disperses in polar aprotic solvents. Aedotron C3-NM is a triblock copolymer.
Please follow this link for Aedotron/Oligotron selection guide table.

Application

Conducting polymer dispersion suitable for making conducting films, anti-static layers and transparent electrodes for electroluminescent lamps.
Nitromethane dispersions can be used to cast thin films on glass, ITO, silicon wafers and many polymeric substrates. Compatibility of the polymeric substrate with nitromethane should be tested in each case. Coat the substrate with a "pool" of the nitromethane dispersion. Thin transparent films are typically spun at 1000 RPM or higher speeds, but more conducting films can be obtained at lower speeds. A single layer spun at 1000 RPM is typically 75 nm thick.

Features and Benefits

The copolymer structure renders the conducting polymer PEDOT highly dispersible in organic solvents. Nitromethane has a suitable volatility for spin coating.

Caution

Store this product at room temperature (do not refrigerate or freeze). Some settling will normally occur. Agitate, sonicate, and filter prior to use. A plug of glass wool in a pipette can be used for easy filtration. The shelf life of this product will vary depending on how it is stored and handled. Product should be discarded if heavy aggregation occurs, or if sonication / filtering are inadequate to produce a good casting solution.

Legal Information

Aedotron is a trademark of TDA Research, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 2 - Repr. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

In the field of organic printable electronics, such as OLEDs and organic photovoltaics (OPVs), improved organic conducting and semiconducting materials are needed. The progress in two fields is reviewed in this article.

Conjugated polymers offer charge transport between inorganic, electrically conducting metals and organic, proton-conducting biological systems.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service