Skip to Content
Merck
All Photos(1)

Key Documents

685232

Sigma-Aldrich

Bis[(R,R,S)-DiazaPhos-SPE]

Synonym(s):

2,2′,2″,2′′′-(1,2-Phenylenebis[(1R,3R)-tetrahydro-5,8-dioxo-1H-[1,2,4]diazaphospholo[1,2-a]pyridazine-2,1,3(3H)-triyl])tetrakis(N-[(1S)-1-phenylethyl])benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C78H72N8O8P2
CAS Number:
Molecular Weight:
1311.40
UNSPSC Code:
12352005
PubChem Substance ID:

optical activity

[α]20/D -82.0°, c = 1 in THF

mp

183-195 °C

SMILES string

C[C@H](NC(=O)c1ccccc1[C@H]2N3N([C@H](P2c4ccccc4P5[C@@H](N6N([C@H]5c7ccccc7C(=O)N[C@@H](C)c8ccccc8)C(=O)CCC6=O)c9ccccc9C(=O)N[C@@H](C)c%10ccccc%10)c%11ccccc%11C(=O)N[C@@H](C)c%12ccccc%12)C(=O)CCC3=O)c%13ccccc%13

InChI

1S/C78H72N8O8P2/c1-49(53-27-9-5-10-28-53)79-71(91)57-35-17-21-39-61(57)75-83-67(87)45-46-68(88)84(83)76(62-40-22-18-36-58(62)72(92)80-50(2)54-29-11-6-12-30-54)95(75)65-43-25-26-44-66(65)96-77(63-41-23-19-37-59(63)73(93)81-51(3)55-31-13-7-14-32-55)85-69(89)47-48-70(90)86(85)78(96)64-42-24-20-38-60(64)74(94)82-52(4)56-33-15-8-16-34-56/h5-44,49-52,75-78H,45-48H2,1-4H3,(H,79,91)(H,80,92)(H,81,93)(H,82,94)/t49-,50-,51-,52-,75+,76+,77+,78?,96?/m0/s1

InChI key

NTWCWWNZWYJTEP-GEDBLHTESA-N

Application

Diazaphospholane Ligands for Catalytic Asymmetric Transformations

Catalyst for:
  • Enantioselective synthesis of α,β-unsaturated aldehydes via hydroformylation of dienes
  • Regioselective and enantioselective hydroformylation of dialkyl acrylamides
  • Asymmetric hydroformylation of vinyl acetate
  • Regioselective and stereoselective hydroformylation of alkenes
Diazaphospholane ligands display excellent levels of conversion and selectivity in Rh-catalyzed asymmetric hydroformylation reactions.

Legal Information

Product is sold for R&D purpose only and use by end user in the manufacture of products of commerce is not permitted. Product is sold in association with DowpharmaSM a business unit of The Dow Chemical Company. U.S. Pat. 7,071,357B.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas P Clark et al.
Journal of the American Chemical Society, 127(14), 5040-5042 (2005-04-07)
Azines made by the reaction of hydrazine with ortho-formylbenzoic acid react with 1,2-diphosphinobenzene and either succinyl chloride or phthaloyl chloride in ca. 30% yield to give rac-bis-3,4-diazaphospholanes bearing benzoic acid groups in the 2 and 5 positions. Condensation of the
Highly regio- and enantioselective asymmetric hydroformylation of olefins mediated by 2,5-disubstituted phospholane ligands.
Alex T Axtell et al.
Angewandte Chemie (International ed. in English), 44(36), 5834-5838 (2005-08-13)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service