Skip to Content
Merck
All Photos(3)

Key Documents

683833

Sigma-Aldrich

2-Ethoxycarbonylphenylboronic acid

≥95%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€93.30

€93.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€93.30

About This Item

Linear Formula:
CH3CH2O2C(C6H4)B(OH)2
CAS Number:
Molecular Weight:
193.99
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

€93.30


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95%

form

powder or crystals

mp

128-136 °C

functional group

ester

SMILES string

CCOC(=O)c1ccccc1B(O)O

InChI

1S/C9H11BO4/c1-2-14-9(11)7-5-3-4-6-8(7)10(12)13/h3-6,12-13H,2H2,1H3

InChI key

QZKVVOXAEBCLPZ-UHFFFAOYSA-N

Application

Reactant for:
  • Synthesis of embelin derivatives via Suzuki-Miyaura reaction, cross metathesis and Wittig olefination
  • Asymmetric cyclopropanation of alkenes with di-Me diazomalonate catalyzed by chiral diene-rhodium complexes
  • Preparation of phenylalanines as selective AMPA- and kainate receptor ligands
  • Preparation of aryl pyrazinones via microwave-assisted palladium-catalyzed arylation of resin supported pyrazinones under simultaneous cooling condition
  • Regioselective preparation of (aryl)hydroxyquinolines via Suzuki-Miyaura cross-coupling with chloro(tosyloxy)quinoline under anhydrous conditions followed by nucleophilic deprotection of the tosyl group

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service