Skip to Content
Merck
All Photos(3)

Key Documents

683787

Sigma-Aldrich

2-(Trifluoromethoxy)phenylboronic acid

≥95%

Synonym(s):

2-(Trifluoromethoxy)benzeneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F3CO(C6H4)B(OH)2
CAS Number:
Molecular Weight:
205.93
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

powder or crystals

functional group

fluoro

SMILES string

OB(O)c1ccccc1OC(F)(F)F

InChI

1S/C7H6BF3O3/c9-7(10,11)14-6-4-2-1-3-5(6)8(12)13/h1-4,12-13H

InChI key

AIJCNTOYZPKURP-UHFFFAOYSA-N

General description

May contain varying amounts of anhydride.

Application

Reactant for:
  • Preparation of imidazo[1,5-a]pyrido[3,2-e]pyrazines and imidazo[1,5-a]quinoxalines as orally active phosphodiesterase 10A inhibitors
  • Suzuki-Miyaura cross-coupling reactions
  • Preparation of 3-(5-arylbenzimidazol-2-yl)-1-oxa-2-azaspiro[4.5]decenes as transient receptor potential melastatin 8 antagonists
  • Preparation of biaryl pyrazole carboxamides as sodium channel blockers for treatment of neuropathic pain
  • Preparation of biaryl oxazoles, imidazoles and thiazoles via condensations as sodium channel blockers and CYP inhibitors for treatment of neuropathic pain

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service