Skip to Content
Merck
All Photos(3)

Key Documents

683388

Sigma-Aldrich

[Pd(acac)2]

Umicore, 99%

Synonym(s):

Palladium(II) acetylacetonate, Diacetylacetonatopalladium(II), Palladium(II) 2,4-pentanedionate, Pd(acac)2

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€46.40
2 G
€134.00

€46.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€46.40
2 G
€134.00

About This Item

Linear Formula:
Pd(C5H7O2)2
CAS Number:
Molecular Weight:
304.64
Beilstein:
4136188
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

€46.40


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

powder

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

200-251 °C (dec.)

SMILES string

CC(=O)\C=C(\C)O[Pd]O\C(C)=C/C(C)=O

InChI

1S/2C5H8O2.Pd/c2*1-4(6)3-5(2)7;/h2*3,6H,1-2H3;/q;;+2/p-2/b2*4-3-;

InChI key

JKDRQYIYVJVOPF-FDGPNNRMSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Precatalysts for asymmetric catalysis offer batch-to-batch consistency in a range of catalytic reactions, supporting diverse research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service