Skip to Content
Merck
All Photos(1)

Key Documents

682977

Sigma-Aldrich

(11bR,11′bR)-4,4′-(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis-dinaphtho[2,1-d:1′, 2′-f][1,3,2]dioxaphosphepin

Synonym(s):

R, R-Reetz X-Diphosphonite

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€118.00

€118.00


Estimated to ship on14 April 2025


Request a Bulk Order

Select a Size

Change View
100 MG
€118.00

About This Item

Empirical Formula (Hill Notation):
C55H36O5P2
CAS Number:
Molecular Weight:
838.82
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

€118.00


Estimated to ship on14 April 2025


Request a Bulk Order

form

solid

Quality Level

optical activity

[α]22/D +270°, c = 1 in chloroform

mp

198-217 °C

SMILES string

CC1(C)c2cccc(P3Oc4ccc5ccccc5c4-c6c(O3)ccc7ccccc67)c2Oc8c(cccc18)P9Oc%10ccc%11ccccc%11c%10-c%12c(O9)ccc%13ccccc%12%13

InChI

1S/C55H36O5P2/c1-55(2)41-21-11-23-47(61-57-43-29-25-33-13-3-7-17-37(33)49(43)50-38-18-8-4-14-34(38)26-30-44(50)58-61)53(41)56-54-42(55)22-12-24-48(54)62-59-45-31-27-35-15-5-9-19-39(35)51(45)52-40-20-10-6-16-36(40)28-32-46(52)60-62/h3-32H,1-2H3

InChI key

DXZANFKSPAISOB-UHFFFAOYSA-N

Application

This diphosphonite ligand has been successfully employed in Noyori-type asymmetric transfer hydrogenation of prochiral ketones.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manfred T Reetz et al.
Journal of the American Chemical Society, 128(4), 1044-1045 (2006-01-26)
A BINOL-derived diphosphonite having a xanthene backbone is an excellent bidentate ligand in Ru-catalyzed 2-propanol-based transfer hydrogenation of aryl/alkyl and alkyl/alkyl ketones (ee = 76-99%). Even notoriously difficult ketones such as isopropyl methyl ketone are reduced with extraordinarily high enantioselectivity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service