Skip to Content
Merck
All Photos(1)

Key Documents

68259

Sigma-Aldrich

Ethyl 3-methyl-4-oxocrotonate

≥97.0% (GC)

Synonym(s):

Ethyl trans-γ-oxosenecioate, Ethyl trans-3-methyl-4-oxo-2-butenoate

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
€295.00

€295.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 ML
€295.00

About This Item

Empirical Formula (Hill Notation):
C7H10O3
CAS Number:
Molecular Weight:
142.15
Beilstein:
1704098
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€295.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥97.0% (GC)

form

liquid

refractive index

n20/D 1.461

bp

195-202 °C (lit.)

density

1.044 g/mL at 20 °C (lit.)

functional group

aldehyde
ester

storage temp.

2-8°C

SMILES string

[H]C(=O)\C(C)=C\C(=O)OCC

InChI

1S/C7H10O3/c1-3-10-7(9)4-6(2)5-8/h4-5H,3H2,1-2H3/b6-4+

InChI key

YLFXEUMFBVGYEL-GQCTYLIASA-N

Other Notes

C5-aldehydic ester useful for the synthesis of isoprenoids by Wittig reaction[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E. Aig et al.
Synthetic Communications, 17, 419-419 (1987)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service