Skip to Content
Merck
All Photos(1)

Key Documents

68249

Sigma-Aldrich

(2S,3S)-trans-3-Methyloxirane-2-methyl 4-nitrobenzoate

≥97.0% (sum of enantiomers, HPLC)

Synonym(s):

(2S,3S)-(−)-3-Methylglycidyl 4-nitrobenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11NO5
CAS Number:
Molecular Weight:
237.21
Beilstein:
6274744
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

Assay

≥97.0% (sum of enantiomers, HPLC)

optical activity

[α]20/D −46±2°, c = 2.5% in chloroform

mp

103-105 °C

storage temp.

2-8°C

SMILES string

C[C@@H]1O[C@H]1COC(=O)c2ccc(cc2)[N+]([O-])=O

Other Notes

Protected epoxy alcohol which can be directly substituted at C-3

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S.Y. Ko et al.
The Journal of Organic Chemistry, 52, 667-667 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service