Skip to Content
Merck
All Photos(1)

Key Documents

682233

Sigma-Aldrich

1,2-Bis(2-methoxyphenyl)-1,1,2,2-tetramethyldisilane

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
((CH3O)C6H4Si(CH3)2)2
CAS Number:
Molecular Weight:
330.57
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

refractive index

n20/D 1.5673

density

1.0297 g/mL at 25 °C

SMILES string

COc1ccccc1[Si](C)(C)[Si](C)(C)c2ccccc2OC

InChI

1S/C18H26O2Si2/c1-19-15-11-7-9-13-17(15)21(3,4)22(5,6)18-14-10-8-12-16(18)20-2/h7-14H,1-6H3

InChI key

GTVBFJHBCZGREK-UHFFFAOYSA-N

Application

The lithium reagent derived from this disilane can be used to install a C-Si bond, easily cleaved by Tamao-Fleming oxidation.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T W Lee et al.
Organic letters, 3(21), 3337-3339 (2001-10-12)
[structure: see text]. The new silyllithium reagent 1 and the corresponding cyanocuprate 3 are readily generated under optimal conditions and are useful for the formation of C-Si bonds. Tamao-Fleming oxidation of such C-Si bonds can be effected under very mild

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service