Skip to Content
Merck
All Photos(1)

Key Documents

68170

Sigma-Aldrich

2-Methyloctane

≥99.0% (GC)

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€308.00

€308.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
€308.00

About This Item

Linear Formula:
CH3(CH2)5CH(CH3)2
CAS Number:
Molecular Weight:
128.26
Beilstein:
1696918
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€308.00


Check Cart for Availability

Request a Bulk Order

Assay

≥99.0% (GC)

refractive index

n20/D 1.403

bp

140-144 °C (lit.)

density

0.714 g/mL at 20 °C (lit.)

SMILES string

CCCCCCC(C)C

InChI

1S/C9H20/c1-4-5-6-7-8-9(2)3/h9H,4-8H2,1-3H3

InChI key

ZUBZATZOEPUUQF-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

78.8 °F - closed cup

Flash Point(C)

26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service