Skip to Content
Merck
All Photos(1)

Key Documents

681687

Sigma-Aldrich

N-Boc-1,2-phenylenediamine

95%

Synonym(s):

(2-Aminophenyl)-carbamic acid tert-butyl ester, Mono-N-Boc-o-phenylenediamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16N2O2
CAS Number:
Molecular Weight:
208.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

109-114 °C

SMILES string

CC(C)(C)OC(=O)Nc1ccccc1N

InChI

1S/C11H16N2O2/c1-11(2,3)15-10(14)13-9-7-5-4-6-8(9)12/h4-7H,12H2,1-3H3,(H,13,14)

InChI key

KCZFBLNQOSFGSH-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the American Chemical Society, 115, 1321-1321 (1993)
Jonathan L Sessler et al.
Journal of the American Chemical Society, 127(32), 11442-11446 (2005-08-11)
The synthesis, characterization, and anion-binding properties of a series of 2,6-diamidopyridine dipyrromethane hybrid macrocycles is presented. As part of this work, a new method for effecting the oxidation of dipyrromethane-based macrocycles in organic solvents has been developed. The macrocyclic frameworks

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service