Skip to Content
Merck
All Photos(1)

Key Documents

680192

Sigma-Aldrich

2,2-Bis((4S)-(–)-4-isopropyloxazoline)propane

96%

Synonym(s):

S-4,5-Dihydro-2-(2-((S)-4,5-dihydro-4-isopropyloxazol-2-yl)propan-2-yl)-4-isopropyloxazole

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€146.00

€146.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€146.00

About This Item

Empirical Formula (Hill Notation):
C15H26N2O2
CAS Number:
Molecular Weight:
266.38
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

€146.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

96%

optical activity

[α]/D -121 to -113°, c = 1% in chloroform

refractive index

n20/D 1.4665

density

0.9864 g/mL at 25 °C

functional group

ether

SMILES string

CC(C)[C@H]1COC(=N1)C(C)(C)C2=N[C@H](CO2)C(C)C

InChI

1S/C15H26N2O2/c1-9(2)11-7-18-13(16-11)15(5,6)14-17-12(8-19-14)10(3)4/h9-12H,7-8H2,1-6H3/t11-,12-/m1/s1

InChI key

XFZUPCITFHSROM-VXGBXAGGSA-N

Application

Bisoxazoline ligand - chiral catalyst for asymetric synthesis.[1]

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organic Syntheses, 83, 97-97 (2005)

Articles

BOX ligand complex enhances catalytic enantioselective aziridination of styrene, with phenyl substituents proving superior over t-Bu groups.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service