Skip to Content
Merck
All Photos(1)

Documents

679100

Sigma-Aldrich

Methyl 4-iodobenzoate

97%

Synonym(s):

4-Iodobenzoic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H4CO2CH3
CAS Number:
Molecular Weight:
262.04
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

112-116 °C

SMILES string

COC(=O)c1ccc(I)cc1

InChI

1S/C8H7IO2/c1-11-8(10)6-2-4-7(9)5-3-6/h2-5H,1H3

InChI key

DYUWQWMXZHDZOR-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Murugesan Rajesh et al.
ChemSusChem, 13(9), 2364-2370 (2020-03-20)
The increasing demand for rechargeable batteries induces the development of greener and better devices. Significant advances have been made in the last decade together with a renewed interest in organic electrode materials. Thus, stable electron-donating organic materials are candidates for
Tan L H Doan et al.
Chemistry, an Asian journal, 10(12), 2660-2668 (2015-08-11)
New Zr(IV)- and Hf(IV)-based metal-organic framework photocatalysts, termed VNU-1 and VNU-2 (where VNU = Vietnam National University), were synthesized and their resulting structures fully characterized. By employing a highly π-conjugated linker, namely 1,4-bis(2-[4-carboxyphenyl]ethynyl)benzene, the optical absorption properties were effectively red-shifted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service