Skip to Content
Merck
All Photos(1)

Key Documents

678554

Sigma-Aldrich

Allenylboronic acid pinacol ester

95%

Synonym(s):

2-Allenyl-4,4,5,5-tetramethyl(1,3,2)-dioxaborolane, 4,4,5,5-Tetramethyl-2-(1,2-propadienyl)-1,3,2-dioxaborolane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15BO2
CAS Number:
Molecular Weight:
166.03
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

refractive index

n20/D 1.459

density

0.9237 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC1(C)OB(OC1(C)C)C=C=C

InChI

1S/C9H15BO2/c1-6-7-10-11-8(2,3)9(4,5)12-10/h7H,1H2,2-5H3

InChI key

CJAOMXUZZONOSD-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

150.1 °F - closed cup

Flash Point(C)

65.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service