Skip to Content
Merck
All Photos(2)

Key Documents

677280

Sigma-Aldrich

sSPhos

Synonym(s):

SPhos (water soluble), Sodium 2′-dicyclohexylphosphino-2,6-dimethoxy-1,1′-biphenyl-3-sulfonate hydrate, [1,1′-Biphenyl]-3-sulfonic acid, 2′-(dicyclohexylphosphino)-2,6-dimethoxy- sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€125.00
2 G
€416.00
10 G
€1,120.00

€125.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€125.00
2 G
€416.00
10 G
€1,120.00

About This Item

Empirical Formula (Hill Notation):
C26H34NaO5PS · xH2O
CAS Number:
Molecular Weight:
512.57 (anhydrous basis)
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

€125.00


Check Cart for Availability

Request a Bulk Order

form

solid

Quality Level

reaction suitability

reaction type: Cross Couplings
reagent type: ligand
reaction type: Suzuki-Miyaura Coupling

functional group

phosphine

SMILES string

O.[Na+].COc1ccc(c(OC)c1-c2ccccc2P(C3CCCCC3)C4CCCCC4)S([O-])(=O)=O

InChI

1S/C26H35O5PS.Na.H2O/c1-30-22-17-18-24(33(27,28)29)26(31-2)25(22)21-15-9-10-16-23(21)32(19-11-5-3-6-12-19)20-13-7-4-8-14-20;;/h9-10,15-20H,3-8,11-14H2,1-2H3,(H,27,28,29);;1H2/q;+1;/p-1

InChI key

MAPQBSXKBDVINV-UHFFFAOYSA-M

Legal Information

Usage subject to US Patents 6307087 and 6395916.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

General catalysts for the Suzuki-Miyaura and Sonogashira coupling reactions of aryl chlorides and for the coupling of challenging substrate combinations in water.
Kevin W Anderson et al.
Angewandte Chemie (International ed. in English), 44(38), 6173-6177 (2005-08-13)
David S Surry et al.
Angewandte Chemie (International ed. in English), 47(34), 6338-6361 (2008-07-30)
Palladium-catalyzed amination reactions of aryl halides have undergone rapid development in the last 12 years, largely driven by the implementation of new classes of ligands. Biaryl phosphanes have proven to provide especially active catalysts in this context. This Review discusses
David S Surry et al.
Chemical science, 2(1), 27-50 (2011-01-01)
Dialkylbiaryl phosphines are a valuable class of ligand for Pd-catalyzed amination reactions and have been applied in a range of contexts. This review attempts to aid the reader in the selection of the best choice of reaction conditions and ligand
Ruben Martin et al.
Accounts of chemical research, 41(11), 1461-1473 (2008-07-16)
The cores of many types of polymers, ligands, natural products, and pharmaceuticals contain biaryl or substituted aromatic structures, and efficient methods of synthesizing these structures are crucial to the work of a broad spectrum of organic chemists. Recently, Pd-catalyzed carbon-carbon

Articles

Buchwald phosphine ligands for C-C, C-N, and C-O bond formation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service