Skip to Content
Merck
All Photos(3)

Key Documents

677191

Sigma-Aldrich

(R)-(+)-α,α-Diphenyl-2-pyrrolidinemethanol trimethylsilyl ether

96%

Synonym(s):

(2R)-2-[Diphenyl[(trimethylsilyl)oxy]methyl]pyrrolidine, (R)-α,α-Diphenylprolinol trimethylsilyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€76.60
1 G
€76.60

€76.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€76.60
1 G
€76.60

About This Item

Empirical Formula (Hill Notation):
C20H27NOSi
CAS Number:
Molecular Weight:
325.52
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

€76.60


Check Cart for Availability

Request a Bulk Order

Assay

96%

form

solid

optical activity

[α]22/D +52±5°, c = 1 in chloroform

refractive index

n20/D 1.5509

density

1.0459 g/mL at 25 °C

functional group

phenyl

storage temp.

2-8°C

SMILES string

C[Si](C)(C)OC([C@H]1CCCN1)(c2ccccc2)c3ccccc3

InChI

1S/C20H27NOSi/c1-23(2,3)22-20(19-15-10-16-21-19,17-11-6-4-7-12-17)18-13-8-5-9-14-18/h4-9,11-14,19,21H,10,15-16H2,1-3H3/t19-/m1/s1

InChI key

RSUHWMSTWSSNOW-LJQANCHMSA-N

General description

(R)-(+)-α,α-Diphenyl-2-pyrrolidinemethanol trimethylsilyl ether is a diarylprolinol silyl ether organocatalyst.[1]

Application

(R)-(+)-α,α-Diphenyl-2-pyrrolidinemethanol trimethylsilyl ether can be used as a catalyst to synthesize:
  • Glycofused tricyclic derivatives, which are used in the development of amyloid β-peptide diagnostic tools.[2]
  • Cyclohexene carbaldehyde derivatives by reacting benzoylnitromethane with aliphatic enals.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organocatalytic Cascade Reaction of Aliphatic Enals and Benzoylnitromethane: Synthesis of Enantioenriched Tetrasubstituted Cyclohexene Carbaldehyde
David Rodriguez FA, et al.
Journal of Chemistry, 2020 (2020)
Synthesis and Preliminary Biological Evaluation of Fluorescent Glycofused Tricyclic Derivatives of Amyloid β-Peptide Ligands
D'Orazio G, et al.
European Journal of Organic Chemistry, 2016(9), 1660-1664 (2016)
The diarylprolinol silyl ether system: a general organocatalyst.
Jensen KL, et al.
Accounts of Chemical Research, 45(2), 248-264 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service