Skip to Content
Merck
All Photos(1)

Documents

674559

Sigma-Aldrich

Methyl 2-chloropyridine-3-carboxylate

97%

Synonym(s):

2-Chloro-3-pyridinecarboxylic acid methyl ester, 2-Chloro-nicotinic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6ClNO2
CAS Number:
Molecular Weight:
171.58
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.537

bp

150 °C/1 mmHg

density

1.314 g/mL at 25 °C

SMILES string

COC(=O)c1cccnc1Cl

InChI

1S/C7H6ClNO2/c1-11-7(10)5-3-2-4-9-6(5)8/h2-4H,1H3

InChI key

MYGAJZBZLONIBZ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service