Skip to Content
Merck
All Photos(1)

Key Documents

67410

Sigma-Aldrich

2,3,5-Tri-O-benzoyl-β-D-ribofuranosyl isothiocyanate

≥98.0%

Synonym(s):

β-D-ribofuranosyl isothiocyanate tribenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H21NO7S
CAS Number:
Molecular Weight:
503.52
Beilstein:
1275438
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0%

SMILES string

O=C(OC[C@H]1O[C@@H](N=C=S)[C@H](OC(=O)c2ccccc2)[C@@H]1OC(=O)c3ccccc3)c4ccccc4

InChI

1S/C27H21NO7S/c29-25(18-10-4-1-5-11-18)32-16-21-22(34-26(30)19-12-6-2-7-13-19)23(24(33-21)28-17-36)35-27(31)20-14-8-3-9-15-20/h1-15,21-24H,16H2/t21-,22-,23-,24-/m1/s1

InChI key

AWVSFHIUQVIIOW-MOUTVQLLSA-N

Other Notes

Building block for the synthesis of nucleosides

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C. Gasch et al.
Tetrahedron Asymmetry, 11, 435-435 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service