Skip to Content
Merck
All Photos(2)

Key Documents

672998

Sigma-Aldrich

4-Benzyloxyphenylmagnesium bromide solution

1.0 M in THF

Synonym(s):

p-Benzyloxyphenylmagnesium bromide, Bromo[4-(phenylmethoxy)phenyl]magnesium

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€493.00

€493.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€493.00

About This Item

Empirical Formula (Hill Notation):
C13H11BrMgO
CAS Number:
Molecular Weight:
287.43
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

€493.00


Check Cart for Availability

Request a Bulk Order

reaction suitability

reaction type: Grignard Reaction

Quality Level

concentration

1.0 M in THF

density

1.033 g/mL at 25 °C

functional group

phenyl

SMILES string

Br[Mg]c1ccc(OCc2ccccc2)cc1

InChI

1S/C13H11O.BrH.Mg/c1-3-7-12(8-4-1)11-14-13-9-5-2-6-10-13;;/h1,3-10H,11H2;1H;/q;;+1/p-1

InChI key

NMSSOXCJYITCNG-UHFFFAOYSA-M

Signal Word

Danger

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 2

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

-6.2 °F

Flash Point(C)

-21.2 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service