Skip to Content
Merck
All Photos(1)

Documents

670111

Sigma-Aldrich

D-Prolinamide

≥99.0% (NT)

Synonym(s):

(R)-Pyrrolidine-2-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10N2O
CAS Number:
Molecular Weight:
114.15
Beilstein:
471499
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (NT)

optical activity

[α]/D +110±3°, c = 2 in ethanol

mp

101-103 °C

SMILES string

NC(=O)[C@H]1CCCN1

InChI

1S/C5H10N2O/c6-5(8)4-2-1-3-7-4/h4,7H,1-3H2,(H2,6,8)/t4-/m1/s1

InChI key

VLJNHYLEOZPXFW-SCSAIBSYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anaëlle Tilborg et al.
ChemistryOpen, 2(5-6), 194-199 (2014-02-20)
Single crystals for two polymorphs of the ammonium carbamate self-derivative salt of prolinamide have been successfully obtained and characterized. Decarbonation of the carbamate salts was monitored by calorimetry, confirming stabilization of the reactive carbonated adducts in the solid state. Sublimation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service