Skip to Content
Merck
All Photos(1)

Key Documents

668591

Sigma-Aldrich

QuadraPure® BZA

400-1100 μm, extent of labeling: 20 mg/g loading, macroporous

Synonym(s):

QuadraPure® Benzylamine

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

product line

QuadraPure®

form

solid

reaction suitability

reaction type: solution phase peptide synthesis
reagent type: catalyst
reagent type: chelator

extent of labeling

20 mg/g loading

particle size

400-1100 μm

functional group

amine

Application

Metal Scavenger: Rh, Pd, Cu, Co, Ni

Legal Information

QuadraPure is a registered trademark of Johnson Matthey Finland Oy

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

QuadraPure Cartridges for Removal of Trace Metal from Reaction Mixtures in Flow
A. Hinchcliffe, et al.
Organic Process Research & Development, 11, 477-481 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service