Skip to Content
Merck
All Photos(1)

Key Documents

668311

Sigma-Aldrich

Pyrrole-2-carbonitrile

96%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€117.00

€117.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€117.00

About This Item

Empirical Formula (Hill Notation):
C5H4N2
CAS Number:
Molecular Weight:
92.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€117.00


Check Cart for Availability

Request a Bulk Order

Assay

96%

refractive index

n20/D 1.5513

bp

92-94 °C/2 mmHg

density

1.081 g/mL at 25 °C

functional group

nitrile

SMILES string

N#Cc1ccc[nH]1

InChI

1S/C5H4N2/c6-4-5-2-1-3-7-5/h1-3,7H

InChI key

BQMPGKPTOHKYHS-UHFFFAOYSA-N

General description

Product is a liquid and not a solid

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seungmin Oh et al.
The journal of physical chemistry. B, 123(39), 8274-8284 (2019-09-11)
Ionic liquids with aprotic heterocyclic anions (AHAs) have been developed for CO2 capture but have been considered for other applications as well. Previously, we have shown that AHA ILs, where the only site for reaction with CO2 is the anion
Quintin R Sheridan et al.
The journal of physical chemistry. B, 120(46), 11951-11960 (2016-10-27)
A combination of X-ray scattering experiments and molecular dynamics simulations were conducted to investigate the structure of ionic liquids (ILs) which chemically bind CO
Guokai Cui et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(14), 5632-5639 (2015-02-18)
A new approach has been developed to improve SO2 sorption by cyano-containing ionic liquids (ILs) through tuning the basicity of ILs and cyano-sulfur interaction. Several kinds of cyano-containing ILs with different basicity were designed, prepared, and used for SO2 capture.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service