Skip to Content
Merck
All Photos(1)

Documents

667501

Sigma-Aldrich

Tantalum nitride etchant

Synonym(s):

Tantalum nitride etch

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161700
NACRES:
NA.23

General description

Tantalum nitride etchant is a high purity etchant systems for precise, clean etching of tantalum, tantalum nitride, and tantalum oxide thin films and resists in electronics applications.

Application

Etchant solution for precise, clean etching of tantalum, tantalum nitride, and tantalum oxide thin films and resists in electronics applications

Features and Benefits

Etch rate 70-80 Å / sec @ 25 C. Use with gold positive or negative photoresist. Rinse with Deionized water.

Preparation Note

  • Etch times vary depending on material type (Ta, TaN, or Ta2O5) and purity.
  • Dilute with water or lower the temperature to achieve a lower etch rate. Adding 1 part water to 2 parts etchant or reducing the temperature 10 °C will approximately reduce the etch rate by 50%.
  • Parts to be etched should be placed in etchant solution with mild to moderate mechanical agitation.
  • Tantalum Etchants contain hydrofluoric acid and will attack silicon oxides, titanium, nickel, aluminum, and chromium.
  • Gold would be the ideal etch stop. You might also succeed with tungsten
  • Rinse with Deionized water.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Oral - Acute Tox. 3 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service