Skip to Content
Merck
All Photos(2)

Key Documents

666599

Sigma-Aldrich

4-Bromo-2-furaldehyde

97%

Synonym(s):

4-Bromofuran-2-carboxaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€86.60

€86.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€86.60

About This Item

Empirical Formula (Hill Notation):
C5H3BrO2
CAS Number:
Molecular Weight:
174.98
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€86.60


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

54-58 °C

functional group

aldehyde
bromo

storage temp.

2-8°C

SMILES string

Brc1coc(C=O)c1

InChI

1S/C5H3BrO2/c6-4-1-5(2-7)8-3-4/h1-3H

InChI key

MRGBBKQOSUHKPF-UHFFFAOYSA-N

Application

Building block for the synthesis of furanyl cyclobutenediones, potent G-protein coupled receptor antagonists.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jianhua Chao et al.
Bioorganic & medicinal chemistry letters, 17(13), 3778-3783 (2007-04-27)
A novel series of cyclobutenedione centered C(4)-alkyl substituted furanyl analogs was developed as potent CXCR2 and CXCR1 antagonists. Compound 16 exhibits potent inhibitory activities against IL-8 binding to the receptors (CXCR2 Ki=1 nM, IC(50)=1.3 nM; CXCR1 Ki=3 nM, IC(50)=7.3 nM)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service