Skip to Content
Merck
All Photos(1)

Key Documents

665371

Sigma-Aldrich

3-Amino-6-chloropyridazine

97%

Synonym(s):

6-Chloro-3-pyridazinamine, 6-Chloropyridazinyl-3-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4ClN3
CAS Number:
Molecular Weight:
129.55
EC Number:
MDL number:

Assay

97%

mp

213-215 °C (dec.)

SMILES string

Nc1ccc(Cl)nn1

InChI

1S/C4H4ClN3/c5-3-1-2-4(6)8-7-3/h1-2H,(H2,6,8)

InChI key

DTXVKPOKPFWSFF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service