Skip to Content
Merck
All Photos(3)

Key Documents

665355

Sigma-Aldrich

(S)-(+)-Benzyl(3,5-dioxa-4-phospha-cyclohepta[2,1-a;3,4-a′]dinaphthalen-4-yl)methylamine

97%

Synonym(s):

(+)-N-Benzyl-N-methyl-dinaphtho[2,1-d:1′,2′]dioxaphosphepin-4-amine, (11bS)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€64.60

€64.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€64.60

About This Item

Empirical Formula (Hill Notation):
C28H22NO2P
CAS Number:
Molecular Weight:
435.45
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

€64.60


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

155-159 °C

SMILES string

CN(Cc1ccccc1)P2Oc3ccc4ccccc4c3-c5c(O2)ccc6ccccc56

InChI

1S/C28H22NO2P/c1-29(19-20-9-3-2-4-10-20)32-30-25-17-15-21-11-5-7-13-23(21)27(25)28-24-14-8-6-12-22(24)16-18-26(28)31-32/h2-18H,19H2,1H3

InChI key

OHGGHRJGBXAKKY-UHFFFAOYSA-N

Application

Phosphoramidite ligand used in a rhodium-catalyzed enantioselective 1,4-addition of arylboronic acids to α,β-unsaturated carbonyl compounds.[1] Also used in asymmetric hydrogenation of N-acylated β-dehydroamino esters to enantiopure β-amino acid derivatives.[2]

Legal Information

Sold under license from DSM for research purposes only.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diego Peña et al.
Journal of the American Chemical Society, 124(49), 14552-14553 (2002-12-06)
New and very easily accessible monodentate phosphoramidite ligands have been developed that lead to excellent ee's and full conversions in the hydrogenation of (E)- and (Z)-beta-dehydroamino acid derivatives with both aliphatic and aromatic side chains. Particularly, two different catalytic systems
Journal of Organometallic Chemistry, 692, 428-428 (2007)

Articles

DSM collaboration offers MonoPhos™ ligands for research, based on the BINOL platform by Feringa and co-workers.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service